Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU812239.1 | - | 1133 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Vitis vinifera |