Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ809943.1 | - | 6 - 7 | 1118 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Vitis vinifera Probe spans exons |