Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD22-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ869893.2 | - | 2 - 3 | 142 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |