Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ901233.1 | - | 1 | 1050 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Both primers and probe map within a single exon |