Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | GQ161924.1 | - | 10 - 11 | 1298 | 66 | |
GQ161925.1 | - | 10 - 11 | 1298 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Vitis vinifera Probe spans exons |