Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001216.1 | NP_001001216.1 | 2 - 3 | 1473 | 101 | |
GenBank mRNA | BC067972.1 | - | 2 - 3 | 1473 | 101 | |
CR760274.1 | - | 2 - 3 | 1358 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |