Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001244.1 | NP_001001244.1 | 19 - 20 | 2330 | 81 | |
GenBank mRNA | BC067305.1 | - | 19 - 20 | 2330 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |