Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004791.1 | NP_001004791.1 | 4 - 5 | 463 | 73 | |
XM_012956698.2 | XP_012812152.1 | 4 - 5 | 584 | 73 | ||
GenBank mRNA | BC074526.1 | - | 4 - 5 | 463 | 73 | |
CR762157.2 | - | 4 - 5 | 434 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |