Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004828.1 | NP_001004828.1 | 3 - 4 | 230 | 92 | |
GenBank mRNA | BC074618.1 | - | 3 - 4 | 230 | 92 | |
CR848614.2 | - | 3 - 4 | 215 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |