Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004829.1 | NP_001004829.1 | 6 - 7 | 846 | 90 | |
XM_012959006.2 | XP_012814460.1 | 6 - 7 | 648 | 90 | ||
XM_012959012.2 | XP_012814466.1 | 6 - 7 | 796 | 90 | ||
XM_018091941.1 | XP_017947430.1 | 6 - 7 | 747 | 90 | ||
GenBank mRNA | BC074619.1 | - | 6 - 7 | 846 | 90 | |
CR761555.2 | - | 5 - 6 | 611 | 90 | ||
CR761725.2 | - | 5 - 6 | 619 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |