Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c19orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004911.1 | NP_001004911.1 | 2 - 3 | 257 | 102 | |
XM_012960834.2 | XP_012816288.1 | 2 - 3 | 474 | 102 | ||
XM_012960835.2 | XP_012816289.1 | 3 - 4 | 572 | 102 | ||
XM_012960836.2 | XP_012816290.1 | 3 - 4 | 446 | 102 | ||
GenBank mRNA | BC075349.1 | - | 2 - 3 | 257 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |