Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004964.1 | NP_001004964.1 | 7 - 8 | 623 | 81 | |
GenBank mRNA | BC075475.1 | - | 7 - 8 | 623 | 81 | |
CR761400.2 | - | 7 - 8 | 643 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |