Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004968.1 | NP_001004968.1 | 4 - 5 | 667 | 110 | |
GenBank mRNA | BC074611.1 | - | 4 - 5 | 474 | 110 | |
BC075480.1 | - | 4 - 5 | 667 | 110 | ||
CR760265.2 | - | 4 - 5 | 414 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |