Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005027.3 | NP_001005027.3 | 3 - 4 | 328 | 71 | |
XM_012959973.2 | XP_012815427.1 | 3 - 4 | 625 | 71 | ||
GenBank mRNA | AL791968.2 | - | 3 - 4 | 328 | 71 | |
BC076709.1 | - | 3 - 4 | 316 | 71 | ||
CU025035.1 | - | 3 - 4 | 297 | 71 |
Gene Symbol: LOC100488176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004910430.3 | XP_004910487.1 | 3 - 4 | 874 | 71 | |
XM_018090819.1 | XP_017946308.1 | 3 - 4 | 336 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |