Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hbd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005089.2 | NP_001005089.1 | 2 - 3 | 373 | 92 | |
GenBank mRNA | BC077014.1 | - | 2 - 3 | 374 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |