Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011012.1 | NP_001011012.1 | 3 - 4 | 437 | 69 | |
GenBank mRNA | BC082725.1 | - | 3 - 4 | 437 | 69 | |
CT025404.2 | - | 3 - 4 | 375 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |