Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: castor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011019.1 | NP_001011019.1 | 2 - 3 | 479 | 94 | |
GenBank mRNA | BC082733.1 | - | 2 - 3 | 479 | 94 | |
CR760105.2 | - | 2 - 3 | 421 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |