Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: timm29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011059.1 | NP_001011059.1 | 1 - 2 | 260 | 57 | |
GenBank mRNA | BC084188.1 | - | 1 - 2 | 260 | 57 | |
CR762195.2 | - | 1 - 2 | 129 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |