Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203521.1 | NP_988852.1 | 4 - 5 | 738 | 81 | |
GenBank mRNA | AY079194.1 | - | 4 - 5 | 738 | 81 | |
CR762022.2 | - | 4 - 5 | 710 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |