Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203552.1 | NP_988883.1 | 10 - 11 | 910 | 78 | |
XM_012958741.2 | XP_012814195.1 | 9 - 10 | 843 | 78 | ||
GenBank mRNA | BC059736.1 | - | 10 - 11 | 910 | 78 | |
CR761659.1 | - | 10 - 11 | 873 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |