Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203638.1 | NP_988969.1 | 2 - 3 | 442 | 59 | |
XM_012970245.1 | XP_012825699.1 | 2 - 3 | 59 | |||
XM_012970246.1 | XP_012825700.1 | 2 - 3 | 59 | |||
XM_012970247.2 | XP_012825701.1 | 2 - 3 | 59 | |||
XM_018097064.1 | XP_017952553.1 | 2 - 3 | 59 | |||
GenBank mRNA | BC061416.1 | - | 2 - 3 | 442 | 59 | |
CR761247.2 | - | 2 - 3 | 485 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |