Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203657.1 | NP_988988.1 | 4 - 5 | 693 | 88 | |
XM_012958135.2 | XP_012813589.1 | 4 - 5 | 639 | 88 | ||
GenBank mRNA | BC061308.1 | - | 4 - 5 | 693 | 88 | |
CT025266.1 | - | 3 - 4 | 551 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |