Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203749.1 | NP_989080.1 | 2 - 3 | 1480 | 84 | |
XM_012962838.2 | XP_012818292.1 | 2 - 3 | 1851 | 84 | ||
GenBank mRNA | AB109108.1 | - | 1 - 2 | 259 | 84 | |
AB109109.1 | - | 2 - 3 | 1071 | 84 | ||
AB109110.1 | - | 2 - 3 | 1751 | 84 | ||
AB109111.1 | - | 2 - 3 | 1480 | 84 | ||
AB109112.1 | - | 1 - 2 | 163 | 84 | ||
BC136147.1 | - | 2 - 3 | 1699 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |