Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203761.1 | NP_989092.1 | 1 - 2 | 190 | 78 | |
GenBank mRNA | BC062500.1 | - | 1 - 2 | 190 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |