Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203775.1 | NP_989106.1 | 2 - 3 | 188 | 81 | |
XM_012970259.2 | XP_012825713.1 | 2 - 3 | 266 | 81 | ||
GenBank mRNA | BC062497.1 | - | 2 - 3 | 188 | 81 | |
CR762193.2 | - | 2 - 3 | 160 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |