Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203853.1 | NP_989184.1 | 3 - 4 | 405 | 65 | |
XM_012964435.2 | XP_012819889.1 | 3 - 4 | 471 | 65 | ||
GenBank mRNA | BC063209.1 | - | 3 - 4 | 405 | 65 | |
CR760309.2 | - | 3 - 4 | 421 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |