Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203854.1 | NP_989185.1 | 4 - 5 | 465 | 73 | |
GenBank mRNA | BC063201.1 | - | 4 - 5 | 465 | 73 | |
CR855728.2 | - | 4 - 5 | 518 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |