Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tktl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203896.1 | NP_989227.1 | 11 - 12 | 1532 | 103 | |
GenBank mRNA | BC063226.1 | - | 11 - 12 | 1532 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |