Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203939.1 | NP_989270.1 | 3 - 4 | 404 | 74 | |
GenBank mRNA | BC064206.1 | - | 3 - 4 | 404 | 74 | |
CR760381.2 | - | 3 - 4 | 381 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |