Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203939.1 | NP_989270.1 | 4 - 5 | 564 | 86 | |
GenBank mRNA | BC064206.1 | - | 4 - 5 | 564 | 86 | |
CR760381.2 | - | 4 - 5 | 541 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |