Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203961.1 | NP_989292.1 | 1 - 2 | 88 | 78 | |
GenBank mRNA | BC063923.1 | - | 1 - 2 | 88 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |