Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203965.1 | NP_989296.1 | 3 - 4 | 581 | 115 | |
GenBank mRNA | BC064229.1 | - | 3 - 4 | 581 | 115 | |
CR760461.2 | - | 3 - 4 | 584 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |