Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204046.1 | NP_989377.1 | 16 - 17 | 2174 | 82 | |
XM_012971610.2 | XP_012827064.1 | 16 - 17 | 2250 | 82 | ||
GenBank mRNA | BC064723.1 | - | 16 - 17 | 2174 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |