Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gchfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213715.1 | NP_998880.1 | 2 - 3 | 149 | 63 | |
XM_018096194.1 | XP_017951683.1 | 3 - 4 | 388 | 63 | ||
GenBank mRNA | BC068218.1 | - | 2 - 3 | 149 | 63 | |
CR848610.2 | - | 2 - 3 | 147 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |