Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pam16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004771.2 | NP_001004771.1 | 3 - 4 | 315 | 92 | |
GenBank mRNA | BC084449.1 | - | 3 - 4 | 315 | 92 | |
CR761515.2 | - | 3 - 4 | 265 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |