Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005440.1 | NP_001005440.1 | 7 - 8 | 939 | 85 | |
XM_018092986.1 | XP_017948475.1 | 7 - 8 | 1034 | 85 | ||
XM_018092987.1 | XP_017948476.1 | 7 - 8 | 927 | 85 | ||
XM_018092988.1 | XP_017948477.1 | 7 - 8 | 927 | 85 | ||
GenBank mRNA | BC074546.1 | - | 7 - 8 | 939 | 85 | |
CR760331.2 | - | 7 - 8 | 917 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |