Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006691.1 | NP_001006692.1 | 2 - 3 | 502 | 101 | |
XM_012959488.2 | XP_012814942.1 | 3 - 4 | 610 | 101 | ||
XM_012959498.2 | XP_012814952.1 | 3 - 4 | 1216 | 101 | ||
XM_012959503.2 | XP_012814957.1 | 3 - 4 | 647 | 101 | ||
XM_012959507.2 | XP_012814961.1 | 4 - 5 | 1166 | 101 | ||
XM_012959516.2 | XP_012814970.1 | 3 - 4 | 545 | 101 | ||
GenBank mRNA | BC075392.1 | - | 2 - 3 | 502 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |