Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006753.1 | NP_001006754.1 | 2 - 3 | 324 | 86 | |
GenBank mRNA | BC075528.1 | - | 2 - 3 | 324 | 86 | |
CT485717.2 | - | 2 - 3 | 332 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |