Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pax3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006775.2 | NP_001006776.2 | 4 - 5 | 692 | 68 | |
XM_012970301.2 | XP_012825755.1 | 4 - 5 | 1263 | 68 | ||
XM_012970302.2 | XP_012825756.1 | 4 - 5 | 1260 | 68 | ||
XM_012970303.2 | XP_012825757.1 | 4 - 5 | 1248 | 68 | ||
XM_012970304.2 | XP_012825758.1 | 4 - 5 | 1263 | 68 | ||
XM_012970306.2 | XP_012825760.1 | 4 - 5 | 1188 | 68 | ||
XM_012970307.2 | XP_012825761.1 | 4 - 5 | 1173 | 68 | ||
GenBank mRNA | BC075577.1 | - | 4 - 5 | 756 | 68 | |
CR855399.2 | - | 4 - 5 | 692 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |