Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006786.1 | NP_001006787.1 | 1 - 2 | 806 | 63 | |
GenBank mRNA | BC075597.1 | - | 1 - 2 | 806 | 63 | |
CR926189.2 | - | 1 - 2 | 539 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |