Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006830.1 | NP_001006831.1 | 3 - 4 | 558 | 79 | |
XM_012970309.1 | XP_012825763.1 | 4 - 5 | 812 | 79 | ||
GenBank mRNA | BC076899.1 | - | 3 - 4 | 558 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |