Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006849.1 | NP_001006850.1 | 4 - 5 | 460 | 93 | |
XM_012962891.1 | XP_012818345.1 | 4 - 5 | 671 | 93 | ||
XM_012962892.1 | XP_012818346.1 | 4 - 5 | 522 | 93 | ||
GenBank mRNA | BC076936.1 | - | 4 - 5 | 460 | 93 | |
CR848219.2 | - | 411 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |