Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006864.1 | NP_001006865.1 | 2 - 3 | 932 | 103 | |
XM_012956765.2 | XP_012812219.1 | 2 - 3 | 1281 | 103 | ||
XM_012956766.2 | XP_012812220.1 | 2 - 3 | 1281 | 103 | ||
XM_012956767.2 | XP_012812221.1 | 2 - 3 | 1282 | 103 | ||
GenBank mRNA | BC076974.1 | - | 2 - 3 | 932 | 103 | |
CT030354.1 | - | 2 - 3 | 863 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |