Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006871.1 | NP_001006872.1 | 5 - 6 | 567 | 83 | |
GenBank mRNA | BC076987.1 | - | 5 - 6 | 567 | 83 | |
CR926398.1 | - | 5 - 6 | 508 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |