Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006915.1 | NP_001006916.1 | 1 - 2 | 494 | 103 | |
GenBank mRNA | BC075431.1 | - | 1 - 2 | 494 | 103 | |
CR848225.2 | - | 1 - 2 | 495 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |