Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007481.1 | NP_001007482.1 | 5 - 6 | 435 | 107 | |
XM_018093846.1 | XP_017949335.1 | 5 - 6 | 458 | 107 | ||
XM_018093847.1 | XP_017949336.1 | 4 - 5 | 401 | 107 | ||
XM_018093848.1 | XP_017949337.1 | 5 - 6 | 458 | 107 | ||
GenBank mRNA | BC079922.1 | - | 5 - 6 | 435 | 107 | |
CT027968.1 | - | 5 - 6 | 435 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |