Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007502.1 | NP_001007503.1 | 3 - 4 | 270 | 80 | |
XM_012960902.2 | XP_012816356.1 | 3 - 4 | 435 | 80 | ||
XM_018089361.1 | XP_017944850.1 | 3 - 4 | 290 | 80 | ||
GenBank mRNA | BC079939.1 | - | 3 - 4 | 270 | 80 | |
CR760189.2 | - | 3 - 4 | 267 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |