Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007857.1 | NP_001007858.1 | 6 - 7 | 928 | 106 | |
XM_012956791.2 | XP_012812245.1 | 7 - 8 | 1041 | 106 | ||
XM_012956792.2 | XP_012812246.1 | 8 - 9 | 942 | 106 | ||
XM_012956793.2 | XP_012812247.1 | 7 - 8 | 911 | 106 | ||
GenBank mRNA | BC079959.1 | - | 6 - 7 | 928 | 106 | |
CR855798.2 | - | 6 - 7 | 922 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |