Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pla2g12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007916.1 | NP_001007917.1 | 1 - 2 | 251 | 80 | |
XM_012966460.2 | XP_012821914.1 | 1 - 2 | 778 | 80 | ||
GenBank mRNA | BC080358.1 | - | 1 - 2 | 251 | 80 | |
CR926443.2 | - | 1 - 2 | 297 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |