Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map1lc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007978.2 | NP_001007979.1 | 1 - 2 | 105 | 70 | |
GenBank mRNA | BC080488.1 | - | 1 - 2 | 107 | 70 | |
CR761386.2 | - | 1 - 2 | 151 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |